Product Image
MP Biomedicals
Bovine Collagen, Sterile
Aqueous solution
Have Questions?
Catalog NumberQuantity
MPB021934922020 mL
Catalog number MPB0219349220
Price (SGD)
542.00
Each
Quantity:
20 mL
Price (SGD)
542.00
Each
Contains 0.3% pure bovine collagen buffered with 0.1% acetate. 95% type I collagen, 5% type III collagen. pH (20 °C): 3.2-3.8.
Specifications
FormulationContains 0.3% pure Bovine Collagen Buffered with 0.1% Acetate; 95% Type I Collagen, 5% Type III Collagen
NameCollagen from Bovine
pH Range1.7 to 2.1
Quantity20 mL
SourceBovine
Storage Requirements4°C
FormSolution
Unit SizeEach