Moved Permanently
The document has moved here.
1000 mm Connecting Leads With Plug And Alligator Clip Black | Contact Us | stage-b2b.fishersci.sg 301 Moved Permanently
Moved Permanently
The document has moved here.
301 Moved Permanently
Moved Permanently
The document has moved here.
1000 mm Connecting Leads With Plug And Alligator Clip
Actual product may vary
1000 mm Connecting Leads With Plug And Alligator Clip
Works well for electrical or laboratory testing applications. The 1000 mm Connecting Leads With Plug And Alligator Clip is a great solution in physics classrooms for assembling circuits quickly.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FISS13986Black
FISS13987Red
Catalog number FISS13986
Price (SGD)
-
Color:
Black
  • 40 in. length (1000 mm)
  • Alligator clip at one end, 4 mm plug on the other end
  • Insulated
  • Great for electrical testing applications
  • Works well in physics classroom and lab settings for assembling circuits quickly
Specifications
Product TypeElectronics Kit
For Use With (Equipment)Great for electrical testing applications
Length (English)40 in.
Length (Metric)1000 mm
MaterialCopper, Plastic
Voltage12 V
ColorBlack
DescriptionInsulated, single
IncludesAlligator clip/4 mm plug
Unit SizeEA
301 Moved Permanently
Moved Permanently
The document has moved here.