▸
Glassware Washers and Cleaners
▸
Washer and Cleaner Accessories
▸
Basket System
Actual product may vary
Basket System
Modular basket system for Elmasonic P, Elmasonic Select, EASY
Have Questions?
Change view
Catalog Number
Dimensions (L x W x H)
Model
FBE1113050
255 x 450 x 135 mm
Basket System Modular Type 300
FBE1108916
105 x 195 x 95 mm
Basket System Modular Type 40
2 Options
Catalog number
FBE1113050
Price (SGD)
153.00
Each
Add to cart
Dimensions (L x W x H):
255 x 450 x 135 mm
Model:
Basket System Modular Type 300
Price (SGD)
153.00
Each
Add to cart
Stainless steel with plastic handles
Ergonomic handles
Autoclavable
Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
Square openings in the side walls for mounting silicone holders
Specifications
Type
Modular Basket System
Capacity (Metric)
10.0 kg
Capacity (English)
22 lb.
For Use With (Equipment)
Fisherbrand Select. Fisherbrand EASY. Elmasonic EASY. Elmasonic P
Dimensions (L x W x H)
255 x 450 x 135 mm
Model
Basket System Modular Type 300