Basket System
Actual product may vary
Basket System
Basket System
Basket System
Basket System
Modular basket system for Elmasonic P, Elmasonic Select, EASY
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)Model
FBE1108916105 x 195 x 95 mmBasket System Modular Type 40
FBE1113050255 x 450 x 135 mmBasket System Modular Type 300
Catalog number FBE1108916
Price (SGD)
110.00
Each
Dimensions (L x W x H):
105 x 195 x 95 mm
Model:
Basket System Modular Type 40
Price (SGD)
110.00
Each
  • Stainless steel with plastic handles
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
TypeModular Basket System
Capacity (Metric)3.0 kg
Capacity (English)6.6 lb.
For Use With (Equipment)Fisherbrand Select. Fisherbrand EASY. Elmasonic EASY. Elmasonic P
Dimensions (L x W x H)105 x 195 x 95 mm
ModelBasket System Modular Type 40
Unit SizeEach