Height Adjustable Floor Stand for Herasafe™ 2030i
Actual product may vary
Height Adjustable Floor Stand for Herasafe™ 2030i
Height Adjustable Floor Stand for Herasafe™ 2030i
Thermo Scientific
Height Adjustable Floor Stand for Herasafe™ 2030i
For use with Herasafe 2030i Biological Safety Cabinets
Have Questions?
Catalog NumberLength (Metric) Exterior
501556891300 mm
Catalog number 50155689
HSN 84149090
Price (SGD)
2,182.00
Each
Length (Metric) Exterior:
1300 mm
Price (SGD)
2,182.00
Each
Height Adjustable Floor Stand in increments of 50mm.(Height range 680-880mm), charcoal grey
  • Adjustment steps of 50mm
  • Charcoal grey
  • Height - 680-880mm
Specifications
DescriptionCharcoal Gray
Product TypeHeight Adjustable Floor Stand
Height (English)26.7 in. to 34.6 in.
Height (Metric)680 to 880 mm
Length (Metric) Exterior1300 mm
For Use With (Equipment)Herasafe 2030i Biological Safety Cabinet size 1.2 m
Depth (Metric)800 mm
Unit SizeEach