AlbumiNZ™ Protease-reduced BSA
Actual product may vary
MP Biomedicals
AlbumiNZ™ Protease-reduced BSA
Shows no detectable enzymatic activity
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
MPB0219989880100 g
MPB02199898911000 g
Catalog number MPB0219989880
Price (SGD)
711.00
Each
Quantity:
100 g
Price (SGD)
711.00
Each
AlbumiNZ™ Protease-reduced BSA is useful for immunoassays where proteases may degrade proteins or where peroxidase or alkaline phospahatase may hydrolyze substrates.
  • Chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality
  • No detectable enzymatic activity
  • Biochem/physiol actions: Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies
Physical characteristics:
  • Form: Off-white to brown powder
  • Purity: ≥97%
  • Total Protein: ≥95%
  • Protease: Reduced
  • pH (4% w/v): 6.5 to 7.5
  • Solubility: Dissolves in ≤20 minutes at 20° to 25°C (20% w/w in water)
  • Moisture: ≤5% w/w
  • ImmunoAssay (EIA) Grade
Certifications: ISO, cGMP, EDQM
Specifications
For Use With (Application)Immunoassay
NameAlbumiNZ™ Protease-reduced BSA
pH Range6.5 to 7.5
Purification MethodChromatography
Quantity100 g
SourceBovine Plasma
Storage Requirements4°C