Moved Permanently
The document has moved here.
Acetonitrile (HPLC), Fisher Chemical 4 L | Contact Us | Thermo Scientific Chemicals 301 Moved Permanently
Moved Permanently
The document has moved here.
301 Moved Permanently
Moved Permanently
The document has moved here.
{"path":"/app/resources/fs/templates/src/header.ejs"}
Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
Thermo Scientific Chemicals
Acetonitrile (HPLC), Fisher Chemical
HPLC mobile phase for analytical and preparative separations
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
Packaging:
Amber Glass Bottle
Catalog number FSKA998-4
Price (SGD)
-
Quantity:
4 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetonitrile
Cleanup SymbolSolvent
Melting Point-46°C
Absorbance0.01 max. at 254nm, 0.02 max. at 220nm, 0.04 max. at 210nm, 0.05 max. at 205nm, 0.07 max. at 200nm, 0.15 max. at 195nm, 1 max. at 190nm, Pass Test
CAS75-05-8
View more
{"path":"/app/resources/fs/templates/src/footer.ejs"} 301 Moved Permanently
Moved Permanently
The document has moved here.