Moved Permanently
The document has moved here.
Methanol Fixative 4 x 250 mL | Contact Us | Eng Scientific | stage-b2b.fishersci.sg 301 Moved Permanently
Moved Permanently
The document has moved here.
301 Moved Permanently
Moved Permanently
The document has moved here.
Methanol Fixative
Actual product may vary
Eng Scientific
Methanol Fixative
Methanol Fixative is for Microbiology, Hematology. Use as fixative prior to staining bloody specimen material or blood culture supematant fluid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FISES59964 x 250 mL
FISES59994 x 3800 mL
Catalog number FISES5996
Price (SGD)
-
Quantity:
4 x 250 mL
  • Methanol preserves the morphology of red blood cells as well as bacteria
For in vitro diagnostic use only.
Specifications
TypeMethanol Fixative
Quantity4 x 250 mL
For Use With (Application)For use as a fixative prior to staining bloody specimen material or blood culture supernatant fluid
Packaging TypeBottle
Stain TypeMethanol
Unit SizePK
301 Moved Permanently
Moved Permanently
The document has moved here.