Moved Permanently
The document has moved here.
Porcelain Desiccator Plates 14 cm | Contact Us | stage-b2b.fishersci.sg 301 Moved Permanently
Moved Permanently
The document has moved here.
301 Moved Permanently
Moved Permanently
The document has moved here.
Porcelain Desiccator Plates
Actual product may vary
Porcelain Desiccator Plates
One large center hole
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) OuterProduct Type
FIS08-641A14 cmDesiccator Plate with 5 mm Perforations
FIS08-641C23 cmDesiccator Plate with 5 mm Perforations
Catalog number FIS08-641A
Price (SGD)
-
Diameter (Metric) Outer:
14 cm
Product Type:
Desiccator Plate with 5 mm Perforations
  • Without feet
  • Glazed on one side
Specifications
Diameter (Metric) Outer14 cm
Product TypeDesiccator Plate with 5 mm Perforations
MaterialPorcelain
For Use With (Equipment)Desiccator
Diameter (English) Outer5.51 in.
Unit SizeEA
301 Moved Permanently
Moved Permanently
The document has moved here.