Shelves for Heratherm™ Ovens and Incubators
Actual product may vary
Thermo Scientific
Shelves for Heratherm™ Ovens and Incubators
Perforated and wire mesh shelves
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDepth (English)Width (English)For Use With (Equipment)Length (English)
5013524420.2 in.20.2 in.Heratherm Adv Prot Oven 750I 400V
5012777114.8 in.14.8 in.Heratherm General Incubator 100L12.9 in.
5012777212.9 in.12.9 in.Heratherm General Incubator 180l12.9 in.
5012777412.9 in.12.9 in.Heratherm Adv Prot Incubator 100l17.2 in.
5012776319.8 in.19.8 in.Heratherm Adv Prot Incubator 180l17.2 in.
5012777719.8 in.19.8 in.Heratherm Adv Prot Incubator 180l17.2 in.
5013524121.06 in.21.06 in.Adv Prot/Gen Incubator 400l20.7 in.
5013524221.06 in.38.74 in.Adv Prot/Gen Incubator 750l
5013524521.06 in.21.06 in.Adv Prot OMH400/Gen Inc IGS400, 400l
5013524621.06 in.21.06 in.Adv Prot OMH750/Gen Inc IGS750, 750l
Catalog number 50135244
HSN 84199090
Price (SGD)
434.00
EA
Depth (English):
20.2 in.
Width (English):
20.2 in.
For Use With (Equipment):
Heratherm Adv Prot Oven 750I 400V
Price (SGD)
434.00
EA
Perforated and wire mesh shelves
Specifications
For Use With (Equipment)Heratherm Adv Prot Oven 750I 400V
TypeShelf
Includes(2) Shelf Supports
Width (English)20.2 in.
Width (Metric)51.3 cm
Depth (English)20.2 in.
Depth (Metric)51.3 cm
Unit SizeEA