Search Fisher Scientific
Get optimal performance and ergonomic comfort from these F1 Multichannel Pipettes. Available in 8-, 12- or 16-channel models; choose from a vast selection of Finntip pipette tips for optimal results.
| Catalog Number | Volume (Metric) | Number of Channels | Color |
|---|---|---|---|
| 4661060N | 10 to 100 μL | 12 | Yellow |
| 4661000N | 1 to 10 μL | 8 | Pink |
| 4661010N | 5 to 50 μL | 8 | Yellow |
| 4661020N | 10 to 100 μL | 8 | Yellow |
| 4661030N | 30 to 300 μL | 8 | Orange |
| 4661040N | 1 to 10 μL | 12 | Pink |
| 4661050N | 5 to 50 μL | 12 | Yellow |
| 4661070N | 30 to 300 μL | 12 | Orange |
| 4661080N | 1 to 10 μL | 16 | Purple |
| 4661090N | 5 to 50 μL | 16 | Turquoise |
Get optimal performance and ergonomics from the Finnpipette™ F1 system, which delivers performance with precision, all in a lightweight design that maximizes user comfort and reduces strain. It's the ideal system for applications using expensive reagents and valuable samples.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >